missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chlorophenyl)ethylboronic acid pinacol ester, 97%

CAS: 1315277-53-2 | C14H20BClO2 | 266.572 g/mol

$70.62 - $70.62

Chemical Identifiers

CAS 1315277-53-2
Molecular Formula C14H20BClO2
Molecular Weight (g/mol) 266.572
MDL Number MFCD12546188
InChI Key WZAJMSPJXKZSTL-UHFFFAOYSA-N
Synonym 2-2-3-chlorophenyl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-chlorophenethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-chlorophenyl ethylboronic acid pinacol ester
PubChem CID 73995113
IUPAC Name 2-[2-(3-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2=CC(=CC=C2)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3046403
View Documents
Thermo Scientific Chemicals
H3046403
1 g
Each for $70.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1315277-53-2
266.572
WZAJMSPJXKZSTL-UHFFFAOYSA-N
73995113
B1(OC(C(O1)(C)C)(C)C)CCC2=CC(=CC=C2)Cl
C14H20BClO2
MFCD12546188
2-2-3-chlorophenyl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-chlorophenethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-chlorophenyl ethylboronic acid pinacol ester
2-[2-(3-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

1315277-53-2
1.4965
MFCD12546188
WZAJMSPJXKZSTL-UHFFFAOYSA-N
2-[2-(3-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
73995113
97%
1 g
C14H20BClO2
2-2-3-chlorophenyl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-chlorophenethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-chlorophenyl ethylboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)CCC2=CC(=CC=C2)Cl
266.572
266.58
2-(3-Chlorophenyl)ethylboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only