missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chlorophenyl)ethylboronic acid pinacol ester, 97%

Catalog No. AAH3046403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3046403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3046403 Supplier Thermo Scientific Chemicals Supplier No. H3046403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1315277-53-2 | C14H20BClO2 | 266.572 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1315277-53-2
Molecular Formula C14H20BClO2
Molecular Weight (g/mol) 266.572
MDL Number MFCD12546188
InChI Key WZAJMSPJXKZSTL-UHFFFAOYSA-N
Synonym 2-2-3-chlorophenyl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-chlorophenethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-chlorophenyl ethylboronic acid pinacol ester
PubChem CID 73995113
IUPAC Name 2-[2-(3-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2=CC(=CC=C2)Cl

Specifications

Quantity 1 g
Refractive Index 1.4965
Formula Weight 266.58
Percent Purity 97%
Chemical Name or Material 2-(3-Chlorophenyl)ethylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only