missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Chloromethyl)piperidino]-6-methylpyrazine, 97%, Thermo Scientific™

Catalog No. CC59024CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC59024CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC59024CB Supplier Thermo Scientific Chemicals Supplier No. CC59024CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 937795-92-1
Molecular Formula C11H16ClN3
Molecular Weight (g/mol) 225.72
MDL Number MFCD09879918
InChI Key DTVYKEHYUMEUFH-UHFFFAOYNA-N
Synonym 2-3-chloromethyl piperidin-1-yl-6-methylpyrazine, 2-3-chloromethyl piperidino-6-methylpyrazine, 2-3-chloromethyl piperidyl-6-methylpyrazine
PubChem CID 24229601
IUPAC Name 2-[3-(chloromethyl)piperidin-1-yl]-6-methylpyrazine
SMILES CC1=CN=CC(=N1)N1CCCC(CCl)C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 225.72
Percent Purity 97%
Chemical Name or Material 2-[3-(Chloromethyl)piperidino]-6-methylpyrazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.