missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Butynyloxy)tetrahydro-2H-pyran, 97%

Catalog No. AC428210050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428210050 5 g Glass bottle
AC428210250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428210050 Supplier Thermo Scientific Chemicals Supplier No. 428210050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40365-61-5 | C9H14O2 | 154.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40365-61-5
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.21
MDL Number MFCD00012352
InChI Key ZQZSNKJFOFAJQX-UHFFFAOYSA-N
Synonym 2-but-3-yn-1-yloxy tetrahydro-2h-pyran, 2-3-butynyloxy tetrahydro-2h-pyran, 2h-pyran, 2-3-butynyloxy tetrahydro, 2-3-butynyloxy tetrahydro-2 h-pyran, 1-tetrahydropyran-2-yloxy-3-butyne, 2-but-3-ynoxytetrahydropyran, tetrahydropyranyloxy-3-butyne, 2-but-3-yn-1-yloxy oxane, 2-3-butynyloxy tetrahydropyran, 4-tetrahydropyranyl oxy butyne
PubChem CID 142440
IUPAC Name 2-but-3-ynoxyoxane
SMILES C#CCCOC1CCCCO1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 0.9840g/mL
Boiling Point 92.0°C to 95.0°C (18.0 mmHg)
Flash Point 73°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.984
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 2-(3-Butynyloxy)tetrahydro-2H-pyran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only