missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanediol (mixture of stereoisomers) 97.0+%, TCI America™

Catalog No. B068125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B068125G 25 g
B0681100G 100 g
B0681500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B068125G Supplier TCI America Supplier No. B068125G

Chemical Identifiers

CAS 513-85-9
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00004523
InChI Key OWBTYPJTUOEWEK-UHFFFAOYSA-N
Synonym 2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-.
PubChem CID 262
ChEBI CHEBI:62064
IUPAC Name butane-2,3-diol
SMILES CC(C(C)O)O

Specifications

Color Yellow
Boiling Point 182°C
Quantity 25 g
Formula Weight 90.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Butanediol (mixture of stereoisomers)
EINECSNumber (2)-0235
RTECSNumber EK0532000
TSCA Yes