missing translation for 'onlineSavingsMsg'
Learn More

(±)-2,3-Butanediol, 98%

Catalog No. p-7057049
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6636922 100 g
AAH6636936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6636922 Supplier Thermo Scientific Chemicals Supplier No. H6636922

CAS: 513-85-9 | C4H10O2 | 90.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 513-85-9
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00004523
InChI Key OWBTYPJTUOEWEK-UHFFFAOYSA-N
Synonym 2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-.
PubChem CID 262
ChEBI CHEBI:62064
IUPAC Name butane-2,3-diol
SMILES CC(C(C)O)O

Specifications

Melting Point 25°C
Color Colorless to Yellow
Density 1.002 g/mL
Boiling Point 183°C to 184°C
Flash Point 85°C (185°F)
Odor Odorless
Refractive Index 1.433
Quantity 100 g
Beilstein 969165
Sensitivity Hygroscopic
Formula Weight 90.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (±)-2,3-Butanediol
Show More Show Less
Hazard Category H227-H320
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P305+P351+P338-P374-P501c
EINECSNumber 208-173-6
RTECSNumber EK0532000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only