missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%

Catalog No. AC433930010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433930010 1 g Glass bottle
AC433930050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433930010 Supplier Thermo Scientific Chemicals Supplier No. 433930010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 124215-44-7 | C9H18BBrO2 | 248.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124215-44-7
Molecular Formula C9H18BBrO2
Molecular Weight (g/mol) 248.96
MDL Number MFCD10567053
InChI Key CHQXFJUKMDJWHO-UHFFFAOYSA-N
Synonym 2-3-bromopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromopropylboronic acid pinacol ester, 1,3,2-dioxaborolane,2-3-bromopropyl-4,4,5,5-tetramethyl, acmc-1c5j4, 3-bromopropylboronic acid, pinacol ester, 2-3-bromopropyl-4?4?5?5-tetramethyl-1?3?2-dioxaborolane
PubChem CID 14836669
IUPAC Name 2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(CCCBr)OC1(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1770g/mL
Boiling Point 61°C to 64°C (0.4 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.177
Formula Weight 248.95
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only