missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromopropyl)-1,3-dioxolane 97.0+%, TCI America™

Catalog No. B34631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34631G 1 g
B34635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34631G Supplier TCI America Supplier No. B34631G
Only null left

Chemical Identifiers

CAS 62563-07-9
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
InChI Key IQIXIJRPYSOGPY-UHFFFAOYSA-N
Synonym 4-Bromobutyraldehyde Ethylene Acetal
PubChem CID 357783
IUPAC Name 2-(3-bromopropyl)-1,3-dioxolane
SMILES C1COC(O1)CCCBr

Specifications

Color Yellow
Boiling Point 88°C
Quantity 1 g
Formula Weight 195.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Bromopropyl)-1,3-dioxolane
TSCA No
Recommended Storage Refrigerator