missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)ethylamine 95.0+%, TCI America™

Catalog No. B39475G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B39475G Supplier TCI America Supplier No. B39475G

Chemical Identifiers

CAS 58971-11-2
Molecular Formula C8H11BrN
Molecular Weight (g/mol) 201.09
MDL Number MFCD01310790
InChI Key ORHRHMLEFQBHND-UHFFFAOYSA-O
Synonym 3-bromophenethylamine, 2-3-bromophenyl ethanamine, 2-3-bromo-phenyl-ethylamine, 2-3-bromophenyl ethylamine, benzeneethanamine, 3-bromo, m-bromophenethylamine, 2-3-bromophenyl ethan-1-amine, 3-bromobenzeneethanamine, 3-bromophenethylaime, 3-bromobenzeneethylamine
PubChem CID 7021736
IUPAC Name 2-(3-bromophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 g
UN Number 2735
Formula Weight 200.08
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(3-Bromophenyl)ethylamine