missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Bromomethyl)phenyl]thiophene, ≥97%, Thermo Scientific™

Catalog No. CC44508CB
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC44508CB 250mg
CC44508DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC44508CB Supplier Thermo Scientific Chemicals Supplier No. CC44508CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85553-44-2
Molecular Formula C11H9BrS
Molecular Weight (g/mol) 253.157
MDL Number MFCD08435867
InChI Key JQQDEBDYUAKNGD-UHFFFAOYSA-N
Synonym 2-3-bromomethyl phenyl thiophene, 3-2-thienyl benzyl bromide, 2-3-bromomethyl phenyl-thiophene
PubChem CID 13150400
IUPAC Name 2-[3-(bromomethyl)phenyl]thiophene
SMILES C1=CC(=CC(=C1)CBr)C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250mg
Formula Weight 253.16
Percent Purity 97%
Chemical Name or Material 2-[3-(Bromomethyl)phenyl]thiophene
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.