missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(3-fluorophenyl)-5-phenyltetrazolium Chloride 98.0+%, TCI America™

Catalog No. B1054100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1054100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1054100MG Supplier TCI America Supplier No. B1054100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 240800-46-8
Molecular Formula C19H13ClF2N4
Molecular Weight (g/mol) 370.788
MDL Number MFCD00059998
InChI Key ADEOAPBIWHHTEL-UHFFFAOYSA-M
PubChem CID 2736029
IUPAC Name 2,3-bis(3-fluorophenyl)-5-phenyltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC(=CC=C3)F)C4=CC(=CC=C4)F.[Cl-]

Specifications

Quantity 100 mg
Formula Weight 370.79
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(3-fluorophenyl)-5-phenyltetrazolium Chloride