missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide 97.0+%, TCI America™

Catalog No. p-7119102
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
100 mg
Catalog No. Quantity
B1535100MG 100 mg
B15351G 1 g
2 options

Catalog No. B1535100MG

Supplier: TCI America B1535100MG

Only null left

Chemical Identifiers

CAS 220191-36-6
Molecular Formula C18H19NO2S2
Molecular Weight (g/mol) 345.48
MDL Number MFCD00142790
InChI Key OHZCQTZIDIVCPI-UHFFFAOYSA-N
PubChem CID 22023748
IUPAC Name 3,4-bis(2,4,5-trimethylthiophen-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
SMILES CC1=C(C)C(=C(C)S1)C1=C(C(=O)NC1=O)C1=C(C)SC(C)=C1C

Specifications

Melting Point 181°C
Quantity 100 mg
Formula Weight 345.48
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide