missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trimethylquinoxaline, 97%

Catalog No. AAB2143203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2143203 1 g
AAB2143206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2143203 Supplier Thermo Scientific Chemicals Supplier No. B2143203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17635-21-1 | C11H12N2 | 172.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17635-21-1
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.23
MDL Number MFCD00047593
InChI Key GQRWKGBOBWHKHP-UHFFFAOYSA-N
Synonym quinoxaline, 2,3,6-trimethyl, quinoxaline,2,3,6-trimethyl, acmc-20aohl, 6-methyl-2,3-dimethylquinoxaline, 2,3,7-trimethylquinoxaline
PubChem CID 87204
IUPAC Name 2,3,6-trimethylquinoxaline
SMILES CC1=CC=C2N=C(C)C(C)=NC2=C1

Specifications

Melting Point 91°C to 94°C
Quantity 1 g
Formula Weight 172.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2,3,6-Trimethylquinoxaline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-618-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only