missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trifluoropyridine, 97%

Catalog No. p-7047635
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1954003 1 g
AAL1954006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1954003 Supplier Thermo Scientific Chemicals Supplier No. L1954003
Only null left

CAS: 3512-18-3 | C5H2F3N | 133.073 g/mol

2,3,6-Trifluoropyridine is used as an intermediate in organic syntheses and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trifluoropyridine is used as an intermediate in organic syntheses and in pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3512-18-3
Molecular Formula C5H2F3N
Molecular Weight (g/mol) 133.073
MDL Number MFCD03001158
InChI Key HRLIANGJWPJFKW-UHFFFAOYSA-N
Synonym pyridine, 2,3,6-trifluoro, 2,3,6-trifluoro-pyridine, pubchem7955, acmc-1cs48, 2,3,6-tris fluoranyl pyridine
PubChem CID 2783288
IUPAC Name 2,3,6-trifluoropyridine
SMILES C1=CC(=NC(=C1F)F)F

Specifications

Density 1.402
Boiling Point 100°C to 102°C
Flash Point 30°C (86°F)
Refractive Index 1.42
Quantity 1 g
UN Number UN1993
Beilstein 1365146
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 133.07
Percent Purity 97%
Chemical Name or Material 2,3,6-Trifluoropyridine
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only