missing translation for 'onlineSavingsMsg'
Learn More

2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-lactosyl bromide

Catalog No. AAJ6638506
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6638506 5 g
AAJ6638509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6638506 Supplier Thermo Scientific Chemicals Supplier No. J6638506

CAS: 4753-07-5 | C26H35BrO17 | 699.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 4753-07-5
Molecular Formula C26H35BrO17
Molecular Weight (g/mol) 699.45
MDL Number MFCD00153593
InChI Key NLFHLQWXGDPOME-UHFFFAOYNA-N
Synonym bromo heptaacetyl-d-lactoside, stabilized with calcium carbonate, bromo heptaacetyl-d-lactoside, 2,3,6,2 ,3 ,4 ,6-hepta-o-acetyl-a-d-cellobiosyl bromide, 2,2',3,3',4',6,6'-hepta-o-acetyl-alpha-d-lactosyl bromide, 2,3,6,2',3',4',6'-hepta-o-acetyl-1-bromo-1-deoxy-alpha-d-lactopyranose min, 2r,3s,4s,5r,6s-3,4,5-tris acetyloxy-6-2r,3r,4s,5r,6r-4,5-bis acetyloxy-2-acetyloxy methyl-6-bromooxan-3-yl oxy oxan-2-yl methyl acetate, 2-o,3-o,6-o-triacetyl-4-o-2-o,3-o,4-o,6-o-tetraacetyl-beta-d-galactopyranosyl-alpha-d-glucopyranosyl bromide, acetobromo-, a-d-lactoside, bromo 4-o-2 ,3,4,6-tetra-o-acetyl-, a-d-galactopyranosyl-2,3,6-tri-o-acetyl-, a-d-glucopyranoside,
PubChem CID 11136238
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-[(2R,3R,4S,5R,6R)-4,5-diacetyloxy-2-(acetyloxymethyl)-6-bromooxan-3-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC2C(OC(C(C2OC(=O)C)OC(=O)C)Br)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Color White
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform
Formula Weight 699.45
Physical Form Powder
Chemical Name or Material 2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-lactosyl bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only