missing translation for 'onlineSavingsMsg'
Learn More

2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide

Catalog No. AAJ6602409
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6602409 10 g
AAJ6602406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6602409 Supplier Thermo Scientific Chemicals Supplier No. J6602409

CAS: 14227-66-8 | C26H35BrO17 | 699.45 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water. Slightly soluble in ethanol

Notes
Hygroscopic, store away from water/moisture. Keep the container tightly closed. Store away from oxidizing agent. Store under dry inert gas.

Chemical Identifiers

CAS 14227-66-8
Molecular Formula C26H35BrO17
Molecular Weight (g/mol) 699.45
MDL Number MFCD00069839
InChI Key NLFHLQWXGDPOME-GLMDSCOTSA-N
Synonym cel br heptaacetate, alpha-d-cellobiosyl bromide heptaacetate, cellobiosyl bromide heptaacetate, acetobromo-d-cellobiose, 2,3,6,2',3',4',6'-hepta-o-acetyl-a-d-cellobiosyl bromide, 2r,3r,4s,5r,6s-2-acetoxymethyl-6-2r,3r,4s,5r,6r-4,5-diacetoxy-2-acetoxymethyl-6-bromotetrahydro-2h-pyran-3-yl oxy tetrahydro-2h-pyran-3,4,5-triyl triacetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-2r,3r,4s,5r,6r-4,5-bis acetyloxy-2-acetyloxy methyl-6-bromooxan-3-yl oxy oxan-2-yl methyl acetate, alpha-d-glucopyranosyl bromide, 4-o-2,3,4,6-tetra-o-acetyl-beta-d-glucopyranosyl-, triacetate
PubChem CID 11039776
SMILES CC(=O)OC[C@H]1OC(Br)C(OC(C)=O)[C@@H](OC(C)=O)C1O[C@@H]1OC(COC(C)=O)[C@@H](OC(C)=O)C(OC(C)=O)[C@H]1OC(C)=O

Specifications

Melting Point 180°C to 190°C
Color White
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water; Slightly soluble in ethanol
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-[(2R,3R,4S,5R,6R)-4,5-diacetyloxy-2-(acetyloxymethyl)-6-bromooxan-3-yl]oxyoxan-2-yl]methyl acetate
Formula Weight 699.45
Physical Form Powder
Chemical Name or Material 2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only