missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylpyridine 98.0+%, TCI America™

Catalog No. p-7136928
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17225G 5 g
T172225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17225G Supplier TCI America Supplier No. T17225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 695-98-7 | C8H11N | 121.183 g/mol

Chemical Identifiers

CAS 695-98-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00126840
InChI Key GFYHSKONPJXCDE-UHFFFAOYSA-N
Synonym 2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f
PubChem CID 12759
IUPAC Name 2,3,5-trimethylpyridine
SMILES CC1=CC(=C(N=C1)C)C

Specifications

Color Yellow
Boiling Point 65°C
Quantity 5 g
Formula Weight 121.18
Percent Purity ≥98.0% (GC)
Grade Laboratory
Physical Form Liquid
Chemical Name or Material 2,3,5-Trimethylpyridine
EINECSNumber (5)-0715
TSCA No