missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylnaphthalene 95.0+%, TCI America™

Catalog No. T0772100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0772100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0772100MG Supplier TCI America Supplier No. T0772100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2245-38-7
Molecular Formula C13H14
Molecular Weight (g/mol) 170.255
MDL Number MFCD00044834
InChI Key JBXULKRNHAQMAS-UHFFFAOYSA-N
Synonym 2,3,5-trimethylnaphthalene, naphthalene, 1,6,7-trimethyl, naphthalene, 2,3,5-trimethyl, unii-4wc055tbj9, naphthalene,1,6,7-trimethyl, 2,5-trimethylnaphthalene, acmc-1cr32, naphthalene,3,5-trimethyl, naphthalene,6,7-trimethyl, naphthalene, 1,6,7-trimethyl-8ci 9ci
PubChem CID 16732
IUPAC Name 1,6,7-trimethylnaphthalene
SMILES CC1=CC=CC2=CC(=C(C=C12)C)C

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 170.26
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3,5-Trimethylnaphthalene