missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trichloropyridine, 98%

Catalog No. p-7055563
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Catalog No. Quantity
AAH6417322 100 g
AAH6417336 500 g
2 options

Catalog No. AAH6417322

Supplier: Thermo Scientific Chemicals H6417322

Only null left

CAS: 16063-70-0 | C5H2Cl3N | 182.428 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 16063-70-0
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.428
MDL Number MFCD00043007
InChI Key CNLIIAKAAMFCJG-UHFFFAOYSA-N
Synonym pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
PubChem CID 27666
IUPAC Name 2,3,5-trichloropyridine
SMILES C1=C(C=NC(=C1Cl)Cl)Cl

Specifications

Melting Point 46°C to 50°C (literature)
Color White to Yellow
Boiling Point 219°C (literature)
Flash Point 113°C (235°F)
Quantity 100 g
Beilstein 119384
Solubility Information Slightly soluble in water.
Formula Weight 182.43
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 2,3,5-Trichloropyridine
Show More Show Less
EINECSNumber 407-270-2
RTECSNumber UU0525000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only