missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trichlorobenzeneboronic acid, 98%

Catalog No. AAL1751103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1751103 1 g
AAL1751106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1751103 Supplier Thermo Scientific Chemicals Supplier No. L1751103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 212779-19-6 | C6H4BCl3O2 | 225.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 212779-19-6
Molecular Formula C6H4BCl3O2
Molecular Weight (g/mol) 225.26
MDL Number MFCD01630856
InChI Key OPBCCRZCYTUJMS-UHFFFAOYSA-N
Synonym 2,3,5-trichlorobenzeneboronic acid, 2,3,5-trichlorophenyl boronic acid, boronic acid, b-2,3,5-trichlorophenyl, b-2,3,5-trichlorophenyl-boronic acid, pubchem14112, acmc-209fiq, 2,3,5-trichlorophenylboronicacid, 2,3,5-trichlorobenzene boronic acid, 2,3,5-trichloro-phenyl boronic acid, boronic acid, 2,3,5-trichlorophenyl
PubChem CID 5106508
IUPAC Name (2,3,5-trichlorophenyl)boronic acid
SMILES OB(O)C1=CC(Cl)=CC(Cl)=C1Cl

Specifications

Melting Point ∼245°C
Quantity 1 g
Formula Weight 225.27
Percent Purity 98%
Chemical Name or Material 2,3,5-Trichlorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only