missing translation for 'onlineSavingsMsg'
Learn More

2',3',5'-Tri-O-acetylguanosine 98.0+%, TCI America™

Catalog No. T26921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26921G 1 g
T26925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26921G Supplier TCI America Supplier No. T26921G

Chemical Identifiers

CAS 6979-94-8
Molecular Formula C16H19N5O8
Molecular Weight (g/mol) 409.36
MDL Number MFCD00057054,MFCD00057054
InChI Key ULXDFYDZZFYGIY-UHFFFAOYNA-N
Synonym 2',3',5'-tri-o-acetylguanosine, triacetylguanosine, 2',3',5'-triacetylguanosine, guanosine, 2',3',5'-triacetate, 2',3',5'-tri-o-acetyl guanosine, 2 ,3 ,5-triacetylguanosine, ulxdfydzzfygiy-sdbhatresa, guanosine 2',3',5'-triacetate, 2r,3r,4r,5r-3,4-bis acetyloxy-5-2-amino-6-oxo-1h-purin-9-yl oxolan-2-yl methyl acetate, guanosine triacetate
PubChem CID 65173
IUPAC Name [3,4-bis(acetyloxy)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(C(OC(C)=O)C1OC(C)=O)N1C=NC2=C1NC(N)=NC2=O

Specifications

Melting Point 213°C
Color White-Yellow
Quantity 1 g
Formula Weight 409.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′,5′-Tri-O-acetylguanosine
TSCA No
Recommended Storage Freezer