missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 321309-43-7
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD02682016
InChI Key JNHKHFADRGJMJS-UHFFFAOYSA-N
Synonym 2-3,5-dimethyl-1h-pyrazol-4-yl benzoic acid, 4-2-carboxyphenyl-3,5-dimethylpyrazole, benzoic acid,2-3,5-dimethyl-1h-pyrazol-4-yl, 2-3,5-dimethylpyrazol-4-yl benzoic acid, 2-3,5-dimethyl-1h-pyrazol-4-yl-benzoic acid
PubChem CID 2776446
IUPAC Name 2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
SMILES CC1=C(C(C)=NN1)C1=CC=CC=C1C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC15701CB
SDS View Documents
Thermo Scientific Chemicals
CC15701CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC15701DA
SDS View Documents
Thermo Scientific Chemicals
CC15701DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

321309-43-7
216.24
JNHKHFADRGJMJS-UHFFFAOYSA-N
2776446
CC1=C(C(C)=NN1)C1=CC=CC=C1C(O)=O
C12H12N2O2
MFCD02682016
2-3,5-dimethyl-1h-pyrazol-4-yl benzoic acid, 4-2-carboxyphenyl-3,5-dimethylpyrazole, benzoic acid,2-3,5-dimethyl-1h-pyrazol-4-yl, 2-3,5-dimethylpyrazol-4-yl benzoic acid, 2-3,5-dimethyl-1h-pyrazol-4-yl-benzoic acid
2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid

Specifications

321309-43-7
100.0
Amber Glass Bottle
MFCD02682016
2-3,5-dimethyl-1h-pyrazol-4-yl benzoic acid, 4-2-carboxyphenyl-3,5-dimethylpyrazole, benzoic acid,2-3,5-dimethyl-1h-pyrazol-4-yl, 2-3,5-dimethylpyrazol-4-yl benzoic acid, 2-3,5-dimethyl-1h-pyrazol-4-yl-benzoic acid
CC1=C(C(C)=NN1)C1=CC=CC=C1C(O)=O
216.24
216.24
2-(3, 5-Dimethyl-1H-pyrazol-4-yl)benzoic acid
96.5
97%
C12H12N2O2
250mg
JNHKHFADRGJMJS-UHFFFAOYSA-N
2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
2776446
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'