missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. p-7128750
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53601G 1 g
D53605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53601G Supplier TCI America Supplier No. D53601G
Only null left

Chemical Identifiers

CAS 365564-07-4
Molecular Formula C14H21BO4
Molecular Weight (g/mol) 264.128
MDL Number MFCD05865191
InChI Key CZYHRTIJLUONKY-UHFFFAOYSA-N
Synonym 3,5-Dimethoxyphenylboronic Acid Pinacol Ester, 1,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
PubChem CID 16217660
IUPAC Name 2-(3,5-dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)OC)OC

Specifications

Melting Point 90°C
Color White
Quantity 1 g
Formula Weight 264.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane