missing translation for 'onlineSavingsMsg'
Learn More

2-[3,5-di(tert-butyl)-4-hydroxyphenyl]acetic acid, Thermo Scientific™

Catalog No. BTB09885EB
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09885EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB09885EB Supplier Thermo Scientific Chemicals Supplier No. BTB09885EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1611-03-6
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.365
InChI Key QLMGIWHWWWXXME-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxyphenylacetic acid, 3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, benzeneacetic acid,3,5-bis 1,1-dimethylethyl-4-hydroxy, acmc-1bvft, 4-hydroxy-3,5-di-t-butylphenylacetic acid, 3,5-di-t-butyl-4-hydroxyphenyl-acetic acid, 4-hydroxy-3,5-di-t-butyl-phenylacetic acid, 3,5-di-tert-butyl-4-hydroxybenzeneacetic acid
PubChem CID 2775110
IUPAC Name 2-(3,5-ditert-butyl-4-hydroxyphenyl)acetic acid
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(=O)O

Specifications

Packaging Amber glass bottle
Quantity 25g
Formula Weight 264.36
Percent Purity 97%
Chemical Name or Material 2-[3, 5-di(tert-butyl)-4-hydroxyphenyl]acetic acid