missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Collidine, 99%

Catalog No. p-7027070
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2353706 5 g
AAB2353714 25 g
AAB2353722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2353706 Supplier Thermo Scientific Chemicals Supplier No. B2353706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 695-98-7 | C8H11N | 121.183 g/mol

2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 695-98-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00126840
InChI Key GFYHSKONPJXCDE-UHFFFAOYSA-N
Synonym 2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f
PubChem CID 12759
IUPAC Name 2,3,5-trimethylpyridine
SMILES CC1=CC(=C(N=C1)C)C

Specifications

Density 0.935
Boiling Point 182°C to 184°C
Flash Point 74°C (165°F)
Odor Unpleasant
Refractive Index 1.5075
Quantity 5 g
UN Number UN2810
Beilstein 108832
Solubility Information Slightly soluble in water.
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2,3,5-Collidine
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 211-786-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only