missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetramethylpyrazine 98.0+%, TCI America™

Catalog No. T097225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T097225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T097225G Supplier TCI America Supplier No. T097225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1124-11-4
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00006146
InChI Key FINHMKGKINIASC-UHFFFAOYSA-N
Synonym tetramethylpyrazine, ligustrazine, pyrazine, tetramethyl, bs factor, tetramethylpyrazin, liqustrazine, tetrapyrazine, 2,3,5,6-tetramethyl pyrazine, chuanxiongzine, ligustizine
PubChem CID 14296
IUPAC Name 2,3,5,6-tetramethylpyrazine
SMILES CC1=C(N=C(C(=N1)C)C)C

Specifications

Melting Point 86°C
Boiling Point 190°C
Quantity 25 g
Formula Weight 136.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetramethylpyrazine
EINECSNumber (5)-0951
RTECSNumber UQ3905000
TSCA Yes