missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetramethylpyrazine, 98+%

Catalog No. AAA1827914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1827914 25 g
AAA1827922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1827914 Supplier Thermo Scientific Chemicals Supplier No. A1827914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1124-11-4 | C8H12N2 | 136.198 g/mol

2,3,5,6-Tetramethylpyrazine is a flavor ingredient of tobacco. It forms cocrystal with 4-hydroxybenzoic acid which exhibits supramolecular synthon polymorphism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5,6-Tetramethylpyrazine is a flavor ingredient of tobacco. It forms cocrystal with 4-hydroxybenzoic acid which exhibits supramolecular synthon polymorphism.

Solubility
Soluble in water (4 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1124-11-4
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00006146
InChI Key FINHMKGKINIASC-UHFFFAOYSA-N
Synonym tetramethylpyrazine, ligustrazine, pyrazine, tetramethyl, bs factor, tetramethylpyrazin, liqustrazine, tetrapyrazine, 2,3,5,6-tetramethyl pyrazine, chuanxiongzine, ligustizine
PubChem CID 14296
IUPAC Name 2,3,5,6-tetramethylpyrazine
SMILES CC1=C(N=C(C(=N1)C)C)C

Specifications

Melting Point 83°C to 86°C
Density 1.08
pH 7
Boiling Point 190°C
Odor Pleasant
Quantity 25 g
Beilstein 113100
Solubility Information Soluble in water (4g/L at 20°C).
Formula Weight 136.2
Percent Purity ≥98%
Chemical Name or Material 2,3,5,6-Tetramethylpyrazine
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 214-391-2
RTECSNumber UQ3905000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only