missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetramethylbenzenesulfonyl chloride, 98%

Catalog No. p-7021112
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1331309 10 g
AAA1331318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1331309 Supplier Thermo Scientific Chemicals Supplier No. A1331309

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60706-63-0 | C10H13ClO2S | 232.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60706-63-0
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.72
MDL Number MFCD00014723
InChI Key ZCXRROBIIMQMHR-UHFFFAOYSA-N
Synonym 2,3,5,6-tetramethylbenzene-1-sulfonyl chloride, 2,3,5,6-tetramethylbenzenesulphonyl chloride, 2,3,5,6-tetramethyl-benzenesulfonyl chloride, chloro 2,3,5,6-tetramethylphenyl sulfone, durylsulphonyl chloride, acmc-1b645, 2,3,5,6-tetramethylbenzenesulfonylchloride, 2,3,5,6-tetramethylphenylsulfonyl chloride, benzenesulfonylchloride, 2,3,5,6-tetramethyl, benzenesulfonyl chloride, 2,3,5,6-tetramethyl-7ci, 9ci
PubChem CID 143740
IUPAC Name 2,3,5,6-tetramethylbenzenesulfonyl chloride
SMILES CC1=CC(C)=C(C)C(=C1C)S(Cl)(=O)=O

Specifications

Melting Point 97°C to 101°C
Quantity 10 g
UN Number UN3261
Beilstein 1876644
Sensitivity Moisture sensitive
Formula Weight 232.73
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetramethylbenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only