missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetramethyl-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. t145725g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
T145725G 25 g
T14575G 5 g
2 options

Catalog No. T145725G

Supplier: TCI America T145725G

Only null left

Chemical Identifiers

CAS 3102-87-2
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00007905
InChI Key WCZNKVPCIFMXEQ-UHFFFAOYSA-N
Synonym 2,3,5,6-tetramethyl-1,4-phenylenediamine, diaminodurene, 2,3,5,6-tetramethyl-p-phenylenediamine, unii-8875rhz1mm, 3,6-diaminodurene, 4-amino-2,3,5,6-tetramethylaniline, tetramethylbenzene-1,4-diamine, 1,4-benzenediamine, 2,3,5,6-tetramethyl, 2,3,5,6-tetramethyl-1,4-benzenediamine, 2,3,5,6-tetramethyl-para-phenylenediamine
PubChem CID 76548
IUPAC Name 2,3,5,6-tetramethylbenzene-1,4-diamine
SMILES CC1=C(C(=C(C(=C1N)C)C)N)C

Specifications

Melting Point 152°C
Color Yellow
Quantity 25 g
Formula Weight 164.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetramethyl-1,4-phenylenediamine
RTECSNumber CZ1599700
TSCA No