missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetramethyl-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. T14575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14575G 5 g
T145725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14575G Supplier TCI America Supplier No. T14575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3102-87-2
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00007905
InChI Key WCZNKVPCIFMXEQ-UHFFFAOYSA-N
Synonym 2,3,5,6-tetramethyl-1,4-phenylenediamine, diaminodurene, 2,3,5,6-tetramethyl-p-phenylenediamine, unii-8875rhz1mm, 3,6-diaminodurene, 4-amino-2,3,5,6-tetramethylaniline, tetramethylbenzene-1,4-diamine, 1,4-benzenediamine, 2,3,5,6-tetramethyl, 2,3,5,6-tetramethyl-1,4-benzenediamine, 2,3,5,6-tetramethyl-para-phenylenediamine
PubChem CID 76548
IUPAC Name 2,3,5,6-tetramethylbenzene-1,4-diamine
SMILES CC1=C(C(=C(C(=C1N)C)C)N)C

Specifications

Melting Point 152°C
Color Yellow
Quantity 5 g
Formula Weight 164.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetramethyl-1,4-phenylenediamine
RTECSNumber CZ1599700
TSCA No