missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol 97.0+%, TCI America™

Catalog No. T19835G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19835G Supplier TCI America Supplier No. T19835G
Only null left

Chemical Identifiers

CAS 2787-79-3
Molecular Formula C7HF7O
Molecular Weight (g/mol) 234.07
MDL Number MFCD00155984
InChI Key HZQGKHUTYHEFBT-UHFFFAOYSA-N
Synonym alpha,alpha,alpha,2,3,5,6-Heptafluoro-p-cresol, Perfluoro-p-cresol, 2,3,5,6-Tetrafluoro-4-hydroxybenzotrifluoride
PubChem CID 2776810
IUPAC Name 2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenol
SMILES OC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 234.07
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol