missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one 98.0+%, TCI America™

Catalog No. p-7136771
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14315G 5 g
T143125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14315G Supplier TCI America Supplier No. T14315G
Only null left

Chemical Identifiers

CAS 95111-49-2
Molecular Formula C7H3Br4NO3
Molecular Weight (g/mol) 468.721
MDL Number MFCD00067043
InChI Key KKWDBDVGMIDKLP-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one, acmc-209ruw, 2,3,5,6-tetrabromo-4-methyl-4-nitrocyclohexa-2,5-dienone, 4-nitro-4-methyl-2,3,5,6-tetrabromo-2,5-cyclohexadien-1-one, 2,3,5,6-tetrabromo-4-methyl-4-nitro-cyclohexa-2,5-dien-1-one
PubChem CID 2724547
IUPAC Name 2,3,5,6-tetrabromo-4-methyl-4-nitrocyclohexa-2,5-dien-1-one
SMILES CC1(C(=C(C(=O)C(=C1Br)Br)Br)Br)[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 468.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one
TSCA No
Recommended Storage Freezer