missing translation for 'onlineSavingsMsg'
Learn More

2,3:5,6-Di-O-isopropylidene-D-mannofuranose 98.0+%, TCI America™

Catalog No. D24475G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D24475G Supplier TCI America Supplier No. D24475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14131-84-1
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00134206
InChI Key JWWCLCNPTZHVLF-WVHJSVDISA-N
Synonym 2,3:5,6-bis-o-1-methylethylidene-, a-d-mannofuranose, 3ar,4s,6r-6-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-ol
PubChem CID 45039045
IUPAC Name (3aR,4S,6R)-6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
SMILES CC1(OCC(O1)C2C3C(C(O2)O)OC(O3)(C)C)C

Specifications

Melting Point 125°C
Color White
Quantity 5 g
Formula Weight 260.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3:5,6-Di-O-isopropylidene-D-mannofuranose