missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethylpentane 99.0+%, TCI America™

Catalog No. T04825ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04825ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T04825ML Supplier TCI America Supplier No. T04825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 565-75-3
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008924
InChI Key RLPGDEORIPLBNF-UHFFFAOYSA-N
Synonym pentane, 2,3,4-trimethyl, unii-woj1qvr6oa, woj1qvr6oa, 2,3,4-trimethyl-pentane, pentane,3,4-trimethyl, acmc-1ao24, pentane,2,3,4-trimethyl, 4-01-00-00446 beilstein handbook reference, 2,3,4-trimethylpentan-3-yl, 2,3,4-trimethylpentane
PubChem CID 11269
IUPAC Name 2,3,4-trimethylpentane
SMILES CC(C)C(C)C(C)C

Specifications

Color Colorless
Boiling Point 112°C
Quantity 5 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4-Trimethylpentane
RTECSNumber SA3321500
TSCA Yes