missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trimethyl-3-pentanol 98.0+%, TCI America™
Supplier: TCI America D243625G
Specifications
| 2,3,4-Trimethyl-3-pentanol | |
| Yellow | |
| C8H18O | |
| 25 g | |
| 2,3,4-trimethyl-3-pentanol, 1,1-diisopropylethanol, 3-pentanol, 2,3,4-trimethyl, diisopropylmethylcarbinol, unii-e7oyt9fj8f, e7oyt9fj8f, fema no. 3903, 2,3,4-trimethyl-3-pentanol fhfi, acmc-1cqn4, 2,3,4-trimethyl-pentan-3-ol | |
| CC(C)C(C)(C(C)C)O | |
| 130.231 | |
| 130.23 | |
| Liquid |
| 3054-92-0 | |
| 78°C | |
| MFCD00048319 | |
| 1987 | |
| PLSMHHUFDLYURK-UHFFFAOYSA-N | |
| 2,3,4-trimethylpentan-3-ol | |
| 520484 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3054-92-0 | |
| 130.231 | |
| PLSMHHUFDLYURK-UHFFFAOYSA-N | |
| 520484 | |
| CC(C)C(C)(C(C)C)O |
| C8H18O | |
| MFCD00048319 | |
| 2,3,4-trimethyl-3-pentanol, 1,1-diisopropylethanol, 3-pentanol, 2,3,4-trimethyl, diisopropylmethylcarbinol, unii-e7oyt9fj8f, e7oyt9fj8f, fema no. 3903, 2,3,4-trimethyl-3-pentanol fhfi, acmc-1cqn4, 2,3,4-trimethyl-pentan-3-ol | |
| 2,3,4-trimethylpentan-3-ol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No