missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trimethyl-2-pentene 98.0+%, TCI America™
$158.44 - $158.44
Chemical Identifiers
| CAS | 565-77-5 |
|---|---|
| Molecular Formula | C8H16 |
| Molecular Weight (g/mol) | 112.216 |
| MDL Number | MFCD00059214 |
| InChI Key | SZFRZEBLZFTODC-UHFFFAOYSA-N |
| Synonym | 2,3,4-trimethyl-2-pentene, 2-pentene, 2,3,4-trimethyl, pentene, trimethyl, acmc-1asqs, 2,4-trimethyl-2-pentene, 2-pentene,3,4-trimethyl, 2-pentene,2,3,4-trimethyl, ch3 2chc ch3 =c ch3 2 |
| PubChem CID | 11270 |
| IUPAC Name | 2,3,4-trimethylpent-2-ene |
| SMILES | CC(C)C(=C(C)C)C |
Chemical Identifiers
| 565-77-5 | |
| 112.216 | |
| SZFRZEBLZFTODC-UHFFFAOYSA-N | |
| 11270 | |
| CC(C)C(=C(C)C)C |
| C8H16 | |
| MFCD00059214 | |
| 2,3,4-trimethyl-2-pentene, 2-pentene, 2,3,4-trimethyl, pentene, trimethyl, acmc-1asqs, 2,4-trimethyl-2-pentene, 2-pentene,3,4-trimethyl, 2-pentene,2,3,4-trimethyl, ch3 2chc ch3 =c ch3 2 | |
| 2,3,4-trimethylpent-2-ene |
Specifications
| 565-77-5 | |
| 116°C | |
| MFCD00059214 | |
| 3295 | |
| SZFRZEBLZFTODC-UHFFFAOYSA-N | |
| 2,3,4-trimethylpent-2-ene | |
| 11270 | |
| ≥98.0% (GC) | |
| 2,3,4-Trimethyl-2-pentene |
| Colorless | |
| C8H16 | |
| 1 mL | |
| 2,3,4-trimethyl-2-pentene, 2-pentene, 2,3,4-trimethyl, pentene, trimethyl, acmc-1asqs, 2,4-trimethyl-2-pentene, 2-pentene,3,4-trimethyl, 2-pentene,2,3,4-trimethyl, ch3 2chc ch3 =c ch3 2 | |
| CC(C)C(=C(C)C)C | |
| 112.216 | |
| 112.22 | |
| Liquid |
Safety and Handling
TSCA : No