missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethyl-2-pentene 98.0+%, TCI America™

Catalog No. T10801ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10801ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10801ML Supplier TCI America Supplier No. T10801ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 565-77-5
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00059214
InChI Key SZFRZEBLZFTODC-UHFFFAOYSA-N
Synonym 2,3,4-trimethyl-2-pentene, 2-pentene, 2,3,4-trimethyl, pentene, trimethyl, acmc-1asqs, 2,4-trimethyl-2-pentene, 2-pentene,3,4-trimethyl, 2-pentene,2,3,4-trimethyl, ch3 2chc ch3 =c ch3 2
PubChem CID 11270
IUPAC Name 2,3,4-trimethylpent-2-ene
SMILES CC(C)C(=C(C)C)C

Specifications

Color Colorless
Boiling Point 116°C
Quantity 1 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4-Trimethyl-2-pentene