missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethoxybenzeneboronic acid, 98%

Catalog No. AAL1983803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1983803 1 g
AAL1983806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1983803 Supplier Thermo Scientific Chemicals Supplier No. L1983803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 118062-05-8 | C9H13BO5 | 212.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118062-05-8
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.008
MDL Number MFCD01318183
InChI Key LDQLSQRFSNMANA-UHFFFAOYSA-N
Synonym 2,3,4-trimethoxyphenyl boronic acid, 2,3,4-trimethoxybenzeneboronic acid, 2,3,4-trimethoxyphenyl boranediol, 2,3,4-trimethoxylphenylboronic acid, boronic acid, 2,3,4-trimethoxyphenyl, pubchem11893, acmc-2099vt, trimethoxyphenyl boronic acid, 2,3,4-trimethoxyphenylboronsaure, ablock ab-12-9176
PubChem CID 2773582
IUPAC Name (2,3,4-trimethoxyphenyl)boronic acid
SMILES B(C1=C(C(=C(C=C1)OC)OC)OC)(O)O

Specifications

Melting Point 74°C to 79°C
Quantity 1 g
Beilstein 4185924
Formula Weight 212.01
Percent Purity 98%
Chemical Name or Material 2,3,4-Trimethoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only