missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trihydroxybenzaldehyde 98.0+%, TCI America™
$55.92 - $162.17
Chemical Identifiers
| CAS | 2144-08-3 |
|---|---|
| Molecular Formula | C7H6O4 |
| Molecular Weight (g/mol) | 154.12 |
| InChI Key | CRPNQSVBEWWHIJ-UHFFFAOYSA-N |
| Synonym | pyrogallol-4-carboxaldehyde, benzaldehyde, 2,3,4-trihydroxy, pyrogallolaldehyd, pubchem8260, 2,4-trihydroxybenzaldehyde, dsstox_cid_31473, dsstox_rid_97358, dsstox_gsid_57684, ksc490m2j, benzaldehyde,3,4-trihydroxy |
| PubChem CID | 75064 |
| IUPAC Name | 2,3,4-trihydroxybenzaldehyde |
| SMILES | C1=CC(=C(C(=C1C=O)O)O)O |
Chemical Identifiers
| 2144-08-3 | |
| 154.12 | |
| pyrogallol-4-carboxaldehyde, benzaldehyde, 2,3,4-trihydroxy, pyrogallolaldehyd, pubchem8260, 2,4-trihydroxybenzaldehyde, dsstox_cid_31473, dsstox_rid_97358, dsstox_gsid_57684, ksc490m2j, benzaldehyde,3,4-trihydroxy | |
| 2,3,4-trihydroxybenzaldehyde |
| C7H6O4 | |
| CRPNQSVBEWWHIJ-UHFFFAOYSA-N | |
| 75064 | |
| C1=CC(=C(C(=C1C=O)O)O)O |
Specifications
| 2144-08-3 | |
| C7H6O4 | |
| pyrogallol-4-carboxaldehyde, benzaldehyde, 2,3,4-trihydroxy, pyrogallolaldehyd, pubchem8260, 2,4-trihydroxybenzaldehyde, dsstox_cid_31473, dsstox_rid_97358, dsstox_gsid_57684, ksc490m2j, benzaldehyde,3,4-trihydroxy | |
| C1=CC(=C(C(=C1C=O)O)O)O | |
| 154.12 | |
| 154.12 | |
| Crystalline Powder |
| 162°C | |
| 5 g | |
| CRPNQSVBEWWHIJ-UHFFFAOYSA-N | |
| 2,3,4-trihydroxybenzaldehyde | |
| 75064 | |
| ≥98.0% (GC,T) | |
| 2,3,4-Trihydroxybenzaldehyde |
Safety and Handling
RTECSNumber : CU8439800
TSCA : Yes