Learn More
2,3,4-Trihydroxybenzaldehyde, 98%
CAS: 2144-08-3 | C8H9BO2 | 147.97 g/mol
$88.99 - $310.70
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1374506
|
Thermo Scientific Chemicals
A1374506 |
5 g |
Each for $88.99
|
|
|||||
|
AAA1374514
|
Thermo Scientific Chemicals
A1374514 |
25 g |
Each for $310.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2144-08-3 |
Specifications
| 2144-08-3 | |
| C8H9BO2 | |
| 5 g | |
| Air Sensitive | |
| QWMJEUJXWVZSAG-UHFFFAOYSA-N | |
| (4-ethenylphenyl)boronic acid | |
| 2734393 | |
| 98% |
| 159°C to 163°C | |
| MFCD00239441 | |
| 2328658 | |
| 4-vinylphenylboronic acid, 4-vinylbenzeneboronic acid, 4-vinylphenyl boronic acid, 4-ethenylphenyl boronic acid, 4-vinylboronic acid, p-styrylboronic acid, p-vinylphenylboronic acid, styrene-4-boronic acid, boronic acid, 4-ethenylphenyl, 4-styreneboronic acid | |
| OB(O)C1=CC=C(C=C)C=C1 | |
| 147.97 | |
| 154.12 | |
| 2,3,4-Trihydroxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 218-404-2
RTECSNumber : CU8439800
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only