missing translation for 'onlineSavingsMsg'
Learn More

2',3',4'-Trihydroxyacetophenone 98.0+%, TCI America™

Catalog No. p-7136654
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11985G 5 g
T119825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11985G Supplier TCI America Supplier No. T11985G
Only null left

Chemical Identifiers

CAS 528-21-2
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00002193
InChI Key XIROXSOOOAZHLL-UHFFFAOYSA-N
Synonym gallacetophenone, 1-2,3,4-trihydroxyphenyl ethanone, 2',3',4'-trihydroxyacetophenone, galloacetophenone, alizarin yellow c, alizarine yellow c, 2,3,4-trihydroxyacetophenone, ethanone, 1-2,3,4-trihydroxyphenyl, 4-acetylpyrogallol, acetophenone, 2',3',4'-trihydroxy
PubChem CID 10706
IUPAC Name 1-(2,3,4-trihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)O)O)O

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′,4′-Trihydroxyacetophenone
RTECSNumber AN0527000
TSCA Yes