Learn More
2,3,4-Trichlorobenzenesulfonyl chloride, 95%
CAS: 34732-09-7 | C6H2Cl4O2S | 279.94 g/mol
$69.56 - $217.14
Chemical Identifiers
| CAS | 34732-09-7 |
|---|---|
| Molecular Formula | C6H2Cl4O2S |
| Molecular Weight (g/mol) | 279.94 |
| MDL Number | MFCD00024871 |
| InChI Key | JDAJYNHGBUXIKS-UHFFFAOYSA-N |
| Synonym | 2,3,4-trichlorobenzene-1-sulfonyl chloride, 2,3,4-trichloro benzene sulfonyl chloride, unii-9qi6w9z13v, benzenesulfonyl chloride, 2,3,4-trichloro, 2,3,4-trichlorobenzenesulfonylchloride, pubchem5173, acmc-1agqg, 2,3,4-trichlorobenzenesulphonyl chloride, jdajynhgbuxiks-uhfffaoysa, chloro 2,3,4-trichlorophenyl sulfone |
| PubChem CID | 36904 |
| IUPAC Name | 2,3,4-trichlorobenzenesulfonyl chloride |
| SMILES | C1=CC(=C(C(=C1S(=O)(=O)Cl)Cl)Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1586503
|
Thermo Scientific Chemicals
A1586503 |
1 g |
Each for $69.56
|
|
|||||
|
AAA1586506
|
Thermo Scientific Chemicals
A1586506 |
5 g |
Each for $217.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 34732-09-7 | |
| 279.94 | |
| JDAJYNHGBUXIKS-UHFFFAOYSA-N | |
| 36904 | |
| C1=CC(=C(C(=C1S(=O)(=O)Cl)Cl)Cl)Cl |
| C6H2Cl4O2S | |
| MFCD00024871 | |
| 2,3,4-trichlorobenzene-1-sulfonyl chloride, 2,3,4-trichloro benzene sulfonyl chloride, unii-9qi6w9z13v, benzenesulfonyl chloride, 2,3,4-trichloro, 2,3,4-trichlorobenzenesulfonylchloride, pubchem5173, acmc-1agqg, 2,3,4-trichlorobenzenesulphonyl chloride, jdajynhgbuxiks-uhfffaoysa, chloro 2,3,4-trichlorophenyl sulfone | |
| 2,3,4-trichlorobenzenesulfonyl chloride |
Specifications
| 34732-09-7 | |
| C6H2Cl4O2S | |
| 1 g | |
| 2649436 | |
| 2,3,4-trichlorobenzene-1-sulfonyl chloride, 2,3,4-trichloro benzene sulfonyl chloride, unii-9qi6w9z13v, benzenesulfonyl chloride, 2,3,4-trichloro, 2,3,4-trichlorobenzenesulfonylchloride, pubchem5173, acmc-1agqg, 2,3,4-trichlorobenzenesulphonyl chloride, jdajynhgbuxiks-uhfffaoysa, chloro 2,3,4-trichlorophenyl sulfone | |
| C1=CC(=C(C(=C1S(=O)(=O)Cl)Cl)Cl)Cl | |
| 279.94 | |
| 279.96 | |
| 2,3,4-Trichlorobenzenesulfonyl chloride |
| 61°C to 65°C | |
| MFCD00024871 | |
| UN3261 | |
| Moisture sensitive | |
| JDAJYNHGBUXIKS-UHFFFAOYSA-N | |
| 2,3,4-trichlorobenzenesulfonyl chloride | |
| 36904 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 252-174-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only