missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trichlorobenzenesulfonyl chloride, 95%

Catalog No. AAA1586506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1586506 5 g
AAA1586503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1586506 Supplier Thermo Scientific Chemicals Supplier No. A1586506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34732-09-7 | C6H2Cl4O2S | 279.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34732-09-7
Molecular Formula C6H2Cl4O2S
Molecular Weight (g/mol) 279.94
MDL Number MFCD00024871
InChI Key JDAJYNHGBUXIKS-UHFFFAOYSA-N
Synonym 2,3,4-trichlorobenzene-1-sulfonyl chloride, 2,3,4-trichloro benzene sulfonyl chloride, unii-9qi6w9z13v, benzenesulfonyl chloride, 2,3,4-trichloro, 2,3,4-trichlorobenzenesulfonylchloride, pubchem5173, acmc-1agqg, 2,3,4-trichlorobenzenesulphonyl chloride, jdajynhgbuxiks-uhfffaoysa, chloro 2,3,4-trichlorophenyl sulfone
PubChem CID 36904
IUPAC Name 2,3,4-trichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1S(=O)(=O)Cl)Cl)Cl)Cl

Specifications

Melting Point 61°C to 65°C
Quantity 5 g
UN Number UN3261
Beilstein 2649436
Sensitivity Moisture sensitive
Formula Weight 279.96
Percent Purity 97%
Chemical Name or Material 2,3,4-Trichlorobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 252-174-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only