missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trichloroaniline 98.0+%, TCI America™

Catalog No. T07985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07985G 5 g
T079825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07985G Supplier TCI America Supplier No. T07985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 634-67-3
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.46
MDL Number MFCD00042819
InChI Key RRJUYQOFOMFVQS-UHFFFAOYSA-N
Synonym aniline, 2,3,4-trichloro, benzenamine, trichloro, benzenamine, 2,3,4-trichloro, unii-84pw2m8b8c, ccris 2397, 2,3,4-trichlorophenylamine, 2,4-trichloroaniline, aniline,3,4-trichloro, acmc-1ax1h, 2,3,4-trichlorobenzenamine
PubChem CID 12464
IUPAC Name 2,3,4-trichloroaniline
SMILES NC1=CC=C(Cl)C(Cl)=C1Cl

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Boiling Point 292°C
Formula Weight 196.46
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,3,4-Trichloroaniline
EINECSNumber (3)-0290
TSCA No