missing translation for 'onlineSavingsMsg'
Learn More

2,3',4'-Trichloroacetophenone

Catalog No. AAB2539714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2539714 25 g
AAB2539706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2539714 Supplier Thermo Scientific Chemicals Supplier No. B2539714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42981-08-8 | C8H5Cl3O | 223.48 g/mol

2,3',4'-Trichloroacetophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3′,4′-Trichloroacetophenone is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 42981-08-8
Molecular Formula C8H5Cl3O
Molecular Weight (g/mol) 223.48
MDL Number MFCD03970382
InChI Key BYTZWANJVUAPNO-UHFFFAOYSA-N
Synonym 2,3',4'-trichloroacetophenone, 2-chloro-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl chloride, 2-chloro-1-3,4-dichloro-phenyl-ethanone, 2-chloro-1-3,4-dichlorophenyl ethan-1-one, 2,3 ,4-trichloroacetophenone, pubchem20021, acmc-1bn8g, alpha,3',4'-trichloroacetophenone, 2-chloranyl-1-3,4-dichlorophenyl ethanone
PubChem CID 316188
SMILES ClCC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 43°C to 45°C
Boiling Point 171°C to 173°C (15 mmHg)
Quantity 25 g
UN Number UN1759
Solubility Information Soluble in methanol.
IUPAC Name 2-chloro-1-(3,4-dichlorophenyl)ethan-1-one
Formula Weight 223.49
Chemical Name or Material 2,3',4'-Trichloroacetophenone
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only