missing translation for 'onlineSavingsMsg'
Learn More

2,3',4'-Trichloroacetophenone 98.0+%, TCI America™

Catalog No. T19655G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19655G Supplier TCI America Supplier No. T19655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42981-08-8
Molecular Formula C8H5Cl3O
Molecular Weight (g/mol) 223.48
MDL Number MFCD03970382
InChI Key BYTZWANJVUAPNO-UHFFFAOYSA-N
Synonym 2,3',4'-trichloroacetophenone, 2-chloro-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl chloride, 2-chloro-1-3,4-dichloro-phenyl-ethanone, 2-chloro-1-3,4-dichlorophenyl ethan-1-one, 2,3 ,4-trichloroacetophenone, pubchem20021, acmc-1bn8g, alpha,3',4'-trichloroacetophenone, 2-chloranyl-1-3,4-dichlorophenyl ethanone
PubChem CID 316188
IUPAC Name 2-chloro-1-(3,4-dichlorophenyl)ethan-1-one
SMILES ClCC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 44°C
Color White
Quantity 5 g
UN Number 1759
Formula Weight 223.48
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3′,4′-Trichloroacetophenone