missing translation for 'onlineSavingsMsg'
Learn More

2-[3,4-(Methylenedioxy)phenyl]ethylamine, 95%

Catalog No. p-7057098
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6644503 1 g
AAH6644506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6644503 Supplier Thermo Scientific Chemicals Supplier No. H6644503
Only null left

CAS: 1484-85-1 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1484-85-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00060509
InChI Key RRIRDPSOCUCGBV-UHFFFAOYSA-N
Synonym homopiperonylamine, 3,4-methylenedioxyphenethylamine, methylenedioxyphenethylamine, 1,3-benzodioxole-5-ethanamine, 2-1,3-benzodioxol-5-yl ethanamine, 2-benzo d 1,3 dioxol-5-yl ethanamine, methylenedioxyphenylethylamine, benzo-1,3-dioxole-5-ethylamine, 3,4-methylenedioxyphenyl ethylamine, phenethylamine, 3,4-methylenedioxy
PubChem CID 73874
IUPAC Name 2-(1,3-benzodioxol-5-yl)ethanamine
SMILES C1OC2=C(O1)C=C(C=C2)CCN

Specifications

Color Colorless to Yellow
Quantity 1 g
Beilstein 383838
Sensitivity Air sensitive
Formula Weight 165.19
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-[3,4-(Methylenedioxy)phenyl]ethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-060-8
RTECSNumber SH9670000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only