missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Epoxycyclohexyl)ethyltrimethoxysilane 97.0+%, TCI America™

Catalog No. E032725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E032725ML 25 mL
E0327500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E032725ML Supplier TCI America Supplier No. E032725ML

Chemical Identifiers

CAS 3388-04-3
Molecular Formula C11H22O4Si
Molecular Weight (g/mol) 246.38
MDL Number MFCD00014485
InChI Key DQZNLOXENNXVAD-UHFFFAOYNA-N
Synonym Trimethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane
PubChem CID 18819
IUPAC Name trimethoxy-[2-(7-oxabicyclo[4.1.0]heptan-4-yl)ethyl]silane
SMILES CO[Si](CCC1CCC2C(C1)O2)(OC)OC

Specifications

Color Colorless
Boiling Point 310°C
Quantity 25 mL
Formula Weight 246.38
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3,4-Epoxycyclohexyl)ethyltrimethoxysilane
EINECSNumber (3)-2647
RTECSNumber VV4000000
TSCA Yes