missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dimethoxybenzamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide 98.0+%, TCI America™

Catalog No. d527525mg
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D527525MG 25 mg
D5275100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D527525MG Supplier TCI America Supplier No. D527525MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 301305-73-7
Molecular Formula C18H20N2O4S
Molecular Weight (g/mol) 360.428
MDL Number MFCD00617269
InChI Key FSPQCTGGIANIJZ-UHFFFAOYSA-N
Synonym flt-3 inhibitor, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxamide, 2-3,4-dimethoxy-benzoylamino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid amide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-benzo b thiophene-3-carboxamide, benzo b thiophene-3-carboxamide,2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro, acmc-20ekhy, d0x7jk, qcr-73
PubChem CID 1048845
IUPAC Name 2-[(3,4-dimethoxybenzoyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
SMILES COC1=C(C=C(C=C1)C(=O)NC2=C(C3=C(S2)CCCC3)C(=O)N)OC

Specifications

Quantity 25 mg
Formula Weight 360.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,4-Dimethoxybenzamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide
TSCA No
Recommended Storage Freezer (-20 °C)