missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3,4-Dimethoxybenzamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide 98.0+%, TCI America™
Supplier: TCI America D5275100MG
Description
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
2-(3,4-Dimethoxybenzamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide | |
C18H20N2O4S | |
100 mg | |
FSPQCTGGIANIJZ-UHFFFAOYSA-N | |
2-[(3,4-dimethoxybenzoyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide | |
1048845 | |
≥98.0% (HPLC) |
301305-73-7 | |
MFCD00617269 | |
flt-3 inhibitor, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxamide, 2-3,4-dimethoxy-benzoylamino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid amide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-benzo b thiophene-3-carboxamide, benzo b thiophene-3-carboxamide,2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro, acmc-20ekhy, d0x7jk, qcr-73 | |
COC1=C(C=C(C=C1)C(=O)NC2=C(C3=C(S2)CCCC3)C(=O)N)OC | |
360.428 | |
360.43 | |
Crystalline Powder |
Chemical Identifiers
301305-73-7 | |
360.428 | |
FSPQCTGGIANIJZ-UHFFFAOYSA-N | |
1048845 | |
COC1=C(C=C(C=C1)C(=O)NC2=C(C3=C(S2)CCCC3)C(=O)N)OC |
C18H20N2O4S | |
MFCD00617269 | |
flt-3 inhibitor, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxamide, 2-3,4-dimethoxy-benzoylamino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid amide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-benzo b thiophene-3-carboxamide, benzo b thiophene-3-carboxamide,2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro, acmc-20ekhy, d0x7jk, qcr-73Show More | |
2-[(3,4-dimethoxybenzoyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamideShow More |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)